Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(-)-Mannitol

D-(-)-Mannitol

Glass bottle for solids, 1 kg
Catalog # ROTH4175.1
Marketsource
Supplier:   Roth Carl
D-(-)-Mannitol
Catalog # ROTH4175.1
Supplier:  Roth Carl
Marketsource
CAS Number:  69-65-8
D-(-)-Mannitol
D-(-)-Mannitol

Glass bottle for solids, 1 kg

CAS Number:  69-65-8
Price
117,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications