Orders LinkContactUsLinkComponent
Adapters, bends
14/23 NS
  LENZ05025001
Adapters, bends
  LENZ05025001
Adapters, bends
Adapters, bends
14/23 NS
23,10 €
1 * 1 ST
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90°. With 2 standard ground cones.
  • Cone size:
    14/23 NS