Express Eingabe
Austria
AuftragsverfolgungKontakt
Test kits, digital titration method
Dissolved oxygen reagent kit: Winkler method, 1 - 10 mg/l, 50 Tests
Catalog # HACH2272200
Supplier:   Hach
Test kits, digital titration method
Catalog # HACH2272200
Supplier:  Hach
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method
Dissolved oxygen reagent kit: Winkler method, 1 - 10 mg/l, 50 Tests
CAS Number:  
Price
195,00 €
Log in for availability
1 * 50 Tests
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • VE:
    50 Tests
  • Bezeichnung:
    Dissolved oxygen reagent kit: Winkler method
  • Messbereich:
    1 - 10 mg/l