Orders LinkContactUsLinkComponent
Cold traps, Type KF | Avantor
Cold traps, Type KF
Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
Artikelnr. KGWA17402
Anbieter:   KGW
Cold traps, Type KF
Artikelnr. KGWA17402
Anbieter:  KGW
Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
Preis
812,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
Spezifikationen
  • Description:
    Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mm
  • Seq:
    07
Accessories
Produktinformationen
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • KGWA1733
    Plastic ring for cold trap, type KF
    Einloggen für Verfügbarkeit
    1 * 1 ST - 67,90 €
Produktinformationen
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • KGWA1741
    Cooling finger S 29-GL
    Einloggen für Verfügbarkeit
    1 * 1 ST - 298,00 €