Draining racks
Draining racks, for 12 test tubes and 11 flasks, 420×170×300 mm
Catalog # HECH49028010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Made of wire, plastic coated.
- Good chemical resistance
- Good temperature resistance
- Easy to clean
Suitable for hanging or free standing test tubes and flasks with a plastic trough.
Specifications
- Description:Draining racks, for 12 test tubes and 11 flasks
- Seq:02
- W×D×H:420×170×300 mm