ContactUsLinkComponent
Quartz glass basin 3F02T4414A in application temperatures of up to 1050°C (maximum 1600°C), translucent, high chemical resistance a nd purity. 1 * 1 items
Catalog # TSLA11209
Quartz glass basin 3F02T4414A in application temperatures of up to 1050°C (maximum 1600°C), translucent, high chemical resistance a nd purity. 1 * 1 items
Catalog # TSLA11209
Frequently Bought Together