Drying tubes, bend
24/29 NS
Catalog # WITG1.660.024
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass 3.3. Angled at 75° with ground joint cone.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.
Specifications
- Cone size:24/29 NS