Orders LinkContactUsLinkComponent
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
2000 ml, 29/32 NS, 19/26 NS
Catalog # WITG0.770.002
Supplier:   witeg Labortechnik
Flasks, round bottom, with three parallel necks
Catalog # WITG0.770.002
Supplier:  witeg Labortechnik
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
2000 ml, 29/32 NS, 19/26 NS
Price
117,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Specifications
  • Capacity:
    2000 ml
  • Side necks:
    19/26 NS
  • Centre neck:
    29/32 NS