Order Entry
Austria
Orders LinkContactUsLinkComponent
 

VWR®, Digital Timer, Three Channels

Digital timer, 3 channels, Black, 0,01 %, 63,5×22,2×86 mm
Catalog # 609-0092
VWR®, Digital Timer, Three Channels
Catalog # 609-0092
Supplier:  
VWR®, Digital Timer, Three Channels
VWR®, Digital Timer, Three Channels

Digital timer, 3 channels, Black, 0,01 %, 63,5×22,2×86 mm

Product Details & Documents

Timing capacity: 19 hours, 59 minutes, 59 seconds.

  • Features: Count up/down, time-in/-out, memory for T1, T2 and T3 channels, alarm, stopwatch and ¹²/₂₄ hour clock
  • Three-line display allows simultaneous or individual countdown alarm timing of up to three tests
  • Loud, high decibel alarm sounds when time is up for 1 minute, or may be silenced manually
  • Extra large digits are easy to read from across the lab

 : Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicating traceability to standards provided by NIST.

 : Supplied with battery, magnetic back and certificate.

Specifications
  • Colour:
    Black
  • Description:
    Digital timer, 3 channels
  • W×D×H:
    63,5×22,2×86 mm
  • Accuracy:
    0,01 %