Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Storage Boxes, Multipurpose

48 l, White, 660×450×220 mm, HDPE
Catalog # BURK3418-0148
Supplier:   BURKLE
Storage Boxes, Multipurpose
Catalog # BURK3418-0148
Supplier:  BURKLE
Storage Boxes, Multipurpose
Storage Boxes, Multipurpose

48 l, White, 660×450×220 mm, HDPE

Price
66,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Made of HDPE, these practical storage containers are ideal for safe transport of pourable or liquid contents.

  • With closed gripping edges
  • Can be stacked
  • Rounded inner edges and smooth inner surfaces for easy cleaning
  • Enables use in foodstuffs/hygiene sector
Specifications
  • Capacity:
    48 l
  • Colour:
    White
  • Material:
    HDPE
  • Seq:
    02
  • W×D×H:
    660×450×220 mm
Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Lid for universal storage containers, PS, white
    Lid for universal storage containers, PS, white
    BURK3418-0001
    - mm
    Log in for availability
    1 * 1 ST - 26,50 €