Orders LinkContactUsLinkComponent
VWR®, Slide Folder
VWR®, Slide Folder
Slide folder, White/black, Cardboard, 20
Catalog # 631-0689
Supplier:   VWR Collection
VWR®, Slide Folder
Catalog # 631-0689
Supplier:  VWR Collection
VWR®, Slide Folder
VWR®, Slide Folder
Slide folder, White/black, Cardboard, 20
Price
11,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Slide trays hold 20 or 30 standard microscopy slides (76×26 mm).
  • Thumb-cut for easy slide removal
  • Horizontal compartments are recessed, protecting the specimens when the cover is closed
Colour: White (black rims)
Specifications
  • No. of slides:
    20
  • Colour:
    White/black
  • Description:
    Slide folder
  • Material:
    Cardboard