Orders LinkContactUsLinkComponent
Percoll™
Percoll™
Percoll™
Catalog # 17-0891-02
Supplier:   Cytiva
Percoll™
Catalog # 17-0891-02
Supplier:  Cytiva
Percoll™
Percoll™
Percoll™
Price
222,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
  • For Research Use Only
  • Commonly used in basic research applications
  • Used in balanced salt solutions
  • Colorless to slightly yellow
A silica-based density gradient media for use in separating cells, organelles, viruses, and other subcellular particles. Percoll® comprises silica particles coated with polyvinylpyrrolidone (PVP) and is commonly used in basic research applications. Also used in balanced salt solutions, physiological saline or 0,25 M sucrose, and for formation of gradients either by the use of convenient gradient mixers or by high speed centrifugation. Colloidal solution of silica coated with polyvinylpyrrolidone (PVP). Liquid/suspension. Colorless to slightly yellow. Certificate of Analysis is available.
Caution: Percoll is for Research use only.
Specifications
  • Description:
    Percoll™
  • Seq:
    01