Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Test tubes, disposable

10 ml, 100 mm, 16 mm
  SEMA2168
 :   SEMADENI
Test tubes, disposable
  SEMA2168
 :  SEMADENI
Test tubes, disposable
Test tubes, disposable

10 ml, 100 mm, 16 mm

 
14,60 €
   
1 * 100 ST
 

PS, clear.

  • Good thermal resistance
  • Good chemical resistance
  • Robust, economical

Universal tubes for a wide range of applications.

 
  • Ext.Ø:
    16 mm
  • Capacity:
    10 ml
  • Length:
    100 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.