TUBING, PTFE, 1/16'' OD X 0.25 MM ID, BL 1 * 10 m
Catalog # JOURJR-T-6808-M10
Supplier: VICI JOUR RESEARCH
TUBING, PTFE, 1/16'' OD X 0.25 MM ID, BL 1 * 10 m
Catalog # JOURJR-T-6808-M10
Supplier: VICI JOUR RESEARCH
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Fine dosage syringes, Injekt®-F Solo
Supplier: B. Braun
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
pH electrodes, SenTix® PLUS
Supplier: WTW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Plastic cuvette for LICO
Supplier: Hach