Orders LinkContactUsLinkComponent
Unions
Union, through bore LP body only, 1/4”-28, for flanged tubing, Polyamide
  JOURJR-060-5
Unions
  JOURJR-060-5
Unions
Union, through bore LP body only, 1/4”-28, for flanged tubing, Polyamide
29,10 €
1 * 5 ST
Unions for easy connection of low pressure solvent lines.
  • Low dead volume
  • Inert and biocompatible
  • Description:
    Union, through bore LP body only, 1/4”-28, for flanged tubing
  • Material:
    Polyamide
  • Seq:
    01