Orders LinkContactUsLinkComponent
Unions
Unions
Union, LP 0,75 mm bore, body only, PEEK
Catalog # JOURJR-068
Supplier:   VICI JOUR RESEARCH
Unions
Catalog # JOURJR-068
Supplier:  VICI JOUR RESEARCH
Unions
Union, LP 0,75 mm bore, body only, PEEK
Price
27,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Unions for easy connection of low pressure solvent lines.
  • Low dead volume
  • Inert and biocompatible
Specifications
  • Description:
    Union, LP 0,75 mm bore, body only
  • Material:
    PEEK
  • Seq:
    03