Order Entry
Austria
Orders LinkContactUsLinkComponent
Nuts with double ferrules

Nuts with double ferrules

Hex-head nut, long
Catalog # JOURJR-5510-5
Supplier:   VICI JOUR RESEARCH
Nuts with double ferrules
Catalog # JOURJR-5510-5
Supplier:  VICI JOUR RESEARCH
Nuts with double ferrules
Nuts with double ferrules

Hex-head nut, long

Price
66,70 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents

These VICI Jour® nuts incorporate a unique PEEK double ferrule which grips the tubing in two places for added security, especially with PEEK or Tefzel tubing. For virtually any ¹/₁₆'' OD fittings eg. Parker, Swagelok, Waters, Valco, Rheodyne, Upchurch.

  • Inert all-PEEK construction
  • Fingertight and hex-head versions
  • Rated up to 420 bar (6000 psi)
Spezifikationen
  • Description:
    Hex-head nut, long
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.