Order Entry
Austria
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Slimsette®, M510

Biopsy processing/embedding cassettes, Slimsette®, M510

Purple
Catalog # 720-1599
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Catalog # 720-1599
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Biopsy processing/embedding cassettes, Slimsette®, M510

Purple

Price
155,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

Acetal polymer. Cassette design is similar to that of the M509 model but specially designed to hold biopsy specimens.

  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Disposable plastic cassettes are highly resistant to histological solutions
  • Small, square openings for good fluid exchange
  • Integral, recessed lid
  • Front face inclined 45° to enable use with certain automatic labelling systems

W×D×H: 41×28,5×6 mm

Packaging: Each case contains three boxes holding 500 cassettes each.

Specifications
  • Colour:
    Purple
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.