Orders LinkContactUsLinkComponent
Slide boxes, SlideFile™ Junior
Slide boxes, SlideFile™ Junior
White, PS, 200, 82×140×86 mm
Catalog # 631-1908
Supplier:   Simport Scientific
Slide boxes, SlideFile™ Junior
Catalog # 631-1908
Supplier:  Simport Scientific
Slide boxes, SlideFile™ Junior
Slide boxes, SlideFile™ Junior
White, PS, 200, 82×140×86 mm
Price
176,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Boxes are impact-resistant and stackable for space efficient storage.
  • Barcodes can be read through the clear lid
  • Number of slides can be doubled by storing two in a slot
  • Tinted hinged cover makes the content easy to see
Ordering information: Include a slide box and a removable tray.
Specifications
  • No. of slides:
    200
  • Colour:
    White
  • Material:
    PS
  • W×D×H:
    82×140×86 mm