Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Slimsette®, M510
Biopsy processing/embedding cassettes, Slimsette®, M510
Grey
Catalog # 720-1612
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Catalog # 720-1612
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Biopsy processing/embedding cassettes, Slimsette®, M510
Grey
Price
155,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
Acetal polymer. Cassette design is similar to that of the M509 model but specially designed to hold biopsy specimens.
  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Disposable plastic cassettes are highly resistant to histological solutions
  • Small, square openings for good fluid exchange
  • Integral, recessed lid
  • Front face inclined 45° to enable use with certain automatic labelling systems
W×D×H: 41×28,5×6 mm
 : Each case contains three boxes holding 500 cassettes each.
Specifications
  • Colour:
    Grey