Chicken Egg White Lysozyme Solution, 10 Mg/Ml 1 * 10 mL
Catalog # 71412-3
Supplier: Merck Millipore (Novagen)
Chicken Egg White Lysozyme Solution, 10 Mg/Ml 1 * 10 mL
Catalog # 71412-3
Supplier: Merck Millipore (Novagen)
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Sampling spoons, sterile, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Accessories for filter plates, 96 well, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
Cell culture vessels, Corning® HYPERflask™ M, CellBIND® treated
Supplier: Corning
VWR®, Microcentrifuge Tubes with Flat Screw Caps
Supplier: VWR Collection
Collagen I-coated plates, Corning® BioCoat®
Supplier: Corning
Bottles, narrow neck, square
Supplier: RIXIUS
Serological pipettes
Supplier: SARSTEDT
CostarRegistered Mark 12 mm TranswellRegistered Mark, 0.4 µm Pore Polyester Membrane Inserts 1 * 48 items
Supplier: STEMCELL Technologies