Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, right-angled, 8,0 mm, 24 NS
Catalog # 189-2124
Supplier:   Lenz Laborglas
Vacuum stopcocks
Catalog # 189-2124
Supplier:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, right-angled, 8,0 mm, 24 NS
Price
85,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifications
  • Hole Ø:
    8,0 mm
  • Description:
    Vacuum stopcocks, one way, right-angled
  • Seq:
    14
  • Spigot:
    24 NS