Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, parallel, 10,0 mm, 29 NS
Catalog # 189-2115
Supplier:   Lenz Laborglas
Vacuum stopcocks
Catalog # 189-2115
Supplier:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, parallel, 10,0 mm, 29 NS
Price
106,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifications
  • Hole Ø:
    10,0 mm
  • Description:
    Vacuum stopcocks, one way, parallel
  • Seq:
    05
  • Spigot:
    29 NS