Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, parallel, 2,5 mm, 14,5 NS
Catalog # 189-2111
Supplier:   Lenz Laborglas
Vacuum stopcocks
Catalog # 189-2111
Supplier:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, parallel, 2,5 mm, 14,5 NS
Price
60,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifications
  • Hole Ø:
    2,5 mm
  • Description:
    Vacuum stopcocks, one way, parallel
  • Seq:
    01
  • Spigot:
    14,5 NS