Order Entry
Austria
Orders LinkContactUsLinkComponent
Nalgene™ Screw Caps

Nalgene™ Screw Caps

Replacement closure, 53 mm, Natural, PP
Catalog # NALG712174-0530
Nalgene™ Screw Caps
Catalog # NALG712174-0530
Supplier:  
Nalgene™ Screw Caps
Nalgene™ Screw Caps

Replacement closure, 53 mm, Natural, PP

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Nalgene® Screw Caps for Teflon Bottles, Tefzel® ETFE, 53-415 GPI 1 * 2 items Catalog #NALG71-2174-0530

Product NALG71-2174-0530 is a replacement for NALG712174-0530 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

PP, coloured.

  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable

Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.

Specifications
  • Cap Size:
    53 mm
  • Colour:
    Natural
  • Description:
    Replacement closure
  • Material:
    PP
  • Seq:
    48
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.