Order Entry
Austria
Orders LinkContactUsLinkComponent
Vortex mixer, VORTEX Genius 3

Vortex mixer, VORTEX Genius 3

VORTEX Genius 3, UK-plug
Catalog # 412-0115
Vortex mixer, VORTEX Genius 3
Catalog # 412-0115
Supplier:  
Vortex mixer, VORTEX Genius 3
Vortex mixer, VORTEX Genius 3

VORTEX Genius 3, UK-plug

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Schüttelgerät Typ Vortex Genius Catalog #412-0098

Product 412-0098 is a replacement for 412-0115 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

Vortex mixer with three interchangeable attachments and various inserts for different applications. Microcentrifuge tubes, microtitre plates, even round bottomed or 250 ml Erlenmeyer flasks can be attached by means of the specially designed strap. Attachments click securely onto the unit in any position. The unit is stable at high speeds and sturdy with coated, zinc die-casting.

  • Compact design
  • Wide speed range, infinitely adjustable
  • Touch or continuous mode
  • Can be used at 5 to 40 °C (80% relative humidity) in an incubator or cold room

IP protection class according to DIN EN 60529: IP 21

* Depending on attachment and loading

Certifications: IP protection class according to DIN EN 60529: IP 21

Ordering information: Supplied with standard attachment for test tubes or small beakers, 412-0099.

Caution: * Depending on attachment and loading
** Use in combination with 412-0101

Specifications
  • Description:
    VORTEX Genius 3, UK-plug
  • Seq:
    02
  • Shaking motion:
    Orbital
  • Cat. No.:
    412-0115
  • Motor input/output:
    58/10 W
  • Orbit:
    4 mm
  • Speed range:
    500 - 2500 min⁻¹*
  • Model:
    VORTEX Genius 3
  • W×D×H:
    127×149×136 mm
  • Weight:
    4,5 kg
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask attachment for Erlenmeyer or round bottom flasks, 100 up to 250 ml**
    Flask attachment for Erlenmeyer or round bottom flasks, 100 up to 250 ml**
    412-0107
    Continuous mode
    Log in for availability
    1 * 1 ST - 43,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube attachment, for 8 reagent tubes, 16 mm**
    Test tube attachment, for 8 reagent tubes, 16 mm**
    412-0105
    Continuous mode
    Log in for availability
    1 * 1 ST - 43,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube attachment, for 18 reagent tubes, 10 mm**
    Test tube attachment, for 18 reagent tubes, 10 mm**
    412-0103
    Continuous mode
    Log in for availability
    1 * 1 ST - 43,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube attachment, for 12 reagent tubes, 12 mm**
    Test tube attachment, for 12 reagent tubes, 12 mm**
    412-0104
    Continuous mode
    Log in for availability
    1 * 1 ST - 43,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube attachment, for 54 microcentrifuge tubes, 1,5/2,0 ml**
    Test tube attachment, for 54 microcentrifuge tubes, 1,5/2,0 ml**
    412-0102
    Continuous mode
    Log in for availability
    1 * 1 ST - 81,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube attachment, for 8 reagent tubes, 20 mm**
    Test tube attachment, for 8 reagent tubes, 20 mm**
    412-0106
    Continuous mode
    Log in for availability
    1 * 1 ST - 43,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Microtitre plate attachment for 1 standard microtitre plate**
    Microtitre plate attachment for 1 standard microtitre plate**
    441-0036
    Continuous mode
    Log in for availability
    1 * 1 ST - 43,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Attachment for single-handed operation, 88 mm round with rubber insert
    Attachment for single-handed operation, 88 mm round with rubber insert
    412-0100
    Continuous or touch mode
    Log in for availability
    1 * 1 ST - 31,50 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Universal attachment, 150 mm with rubber insert
    Universal attachment, 150 mm with rubber insert
    412-0101
    Continuous mode
    Log in for availability
    1 * 1 ST - 35,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Standard attachment
    Standard attachment
    444-2748
    Continuous or touch mode
    Log in for availability
    1 * 1 ST - 14,80 €