Complement reagents
Goat complememnt
ROCKC209-0005
Complement reagents are prepared from healthy, non-medicated animals using very large pools to reduce lot-to-lot variability. The fresh frozen complement is rapidly processed and maintained at –70 °C to ensure maximum activity. The lyophilised complement is supplied with a diluent of veronal buffered saline containing optimal concentrations of magnesium and calcium.
Documents
- Description:Goat complememnt
- Seq:29
- Cat. No.:ROCKC209-0005
- Formulation:Lyophilized