ContactUsLinkComponent
Jetpei-fluor dna transfection reagent is a tetramethylrhodamine-conjugated linear polyethylenimine derivative (excitation at 550 nm emission at 575 nm). 1 * 500 µl
Catalog # 106-05N
undefined
Jetpei-fluor dna transfection reagent is a tetramethylrhodamine-conjugated linear polyethylenimine derivative (excitation at 550 nm emission at 575 nm). 1 * 500 µl
Catalog # 106-05N