Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Ectoine

Glass bottle for solids, 5 g
Catalog # BMOL55459-7
Ectoine
Catalog # BMOL55459-7
CAS Number:  96702-03-3
Ectoine

Glass bottle for solids, 5 g

CAS Number:  96702-03-3
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
  • CAS Number:96702-03-3
  • Formula:C₆H₁₀N₂O₂
  • MDL Number:MFCD03419286
  • Molecular Weight:142.1558 g/mol
  • EINECS:431-910-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications