Formaldehyd 7,5%, TECHNICAL, gepuffert (pH 7,0 ±0,2)
Kunststoffflasche, 1 L
Supplier: VWR Chemicals
Formaldehyd 7,5%, TECHNICAL, gepuffert (pH 7,0 ±0,2)
Catalog # 5534.1000
Supplier: VWR Chemicals
CAS Number: 50-00-0
Springe zu:
Product Details & Documents
Synonyme: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Nummer:50-00-0
- Formel:CH₂O
- MDL-Nummer:MFCD00003274
- Molekulargewicht:30.03 g/mol
- Lagertemperatur:Raumtemperatur
- EINECS:200-001-8
Documents
Zertifikate
Suche nach Chargennummer10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000)
Packaging Information
14543428.pdf 14543429.pdf P20_VWRC5534.1000_SPEC_DE.pdf P20_VWRC5534.9010_SPEC_DE.pdf spec_VWRC5534.1000
Product specification
More
Spezifikationen
Spezifikation Testergebnisse
- Assay (Formaldehyde):7 - 9 %
- pH (20°C):6.8 - 7.2
Spezifikationen
- Packung:Kunststoffflasche
- VE:1 L
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



