Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
20 ml, 10/13, 0,04 ± ml
Catalog # POULRB258/D/5
Supplier:   Poulten & Graf
Volumetric flasks, class A
Catalog # POULRB258/D/5
Supplier:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
20 ml, 10/13, 0,04 ± ml
Price
55,40 €
Log in for availability
1 * 5 ST
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifications
  • NS:
    10/13
  • Capacity:
    20 ml
  • Seq:
    21
  • Tolerance:
    0,04 ± ml