Solid phase microextraction fibers
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specifications
- Description:SPME fiber for autosampler use, carboxen/PDMS, 23 gauge, 75 µm fiber coating
- Seq:16
Accessories
Product Information
- Catalog #AvailabilityPrice
- Merlin microseal, SPME replacement seal, 23 gaugeMerlin microseal, SPME replacement seal, 23 gaugeVARI392609902Log in for availability1 * 1 ST - 439,89 €
Product Information
- Catalog #AvailabilityPrice
- SPME fiber holder for CTC autosampler
Product Information
- Catalog #AvailabilityPrice
- SPME stand, 15 ml
Product Information
- Catalog #AvailabilityPrice
- SPME inlet guide for manual injection