Orders LinkContactUsLinkComponent
Handheld refactometers
Handheld refactometers
30PX, 0 - 85 °Bx ; 1,32 - 1,50 nD, 0,1 °Bx ; ±0,0005 nD, 0,0001 nD
Catalog # 635-0445
Supplier:   Mettler - Toledo
Handheld refactometers
Catalog # 635-0445
Supplier:  Mettler - Toledo
Handheld refactometers
Handheld refactometers
30PX, 0 - 85 °Bx ; 1,32 - 1,50 nD, 0,1 °Bx ; ±0,0005 nD, 0,0001 nD
Price
3 140,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These refractometers, which use the total reflection method, are handheld instruments for measurements in the laboratory and production environments. The backlit LC display and the simple intuitive menu structure allow the user to easily change settings in seconds. The sample can either be placed on the measurement cell or the sensor can be immersed directly into the fluid.
  • Selectable automatic temperature compensation (ATC)
  • Handheld or benchtop instrument
  • Choice of use: Immerse sensor in liquid or add sample to cell
  • Storage of up to 1100 results and easy transfer with IR interface (adapter accessory 635-0117 required)
  • Refracto GS has sapphire prism and thus extended range and faster response
The results can be displayed in a wide variety of standard units and also units applicable to antifreeze and wine measurement as well as customised applications.
Delivery information: Supplied with carry case, HelloCD™ software, two AAA batteries, cleaning tissues, two LDPE pipettes, two PE vials and operating instructions.
Specifications
  • Model:
    30PX
  • Range:
    0 - 85 °Bx ; 1,32 - 1,50 nD
  • Resolution:
    0,0001 nD
  • Precision:
    0,1 °Bx ; ±0,0005 nD
  • Interfaces:
    Infrared for printer or PC
  • Cat. No.:
    635-0445
  • Response time:
    3 - 180 seconds
  • Electrical:
    Two AAA batteries
  • Weight:
    250 g
  • Operating temperature:
    10...40 °C
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Infrared adapter / RS232 DB9M
    Infrared adapter / RS232 DB9M
    635-0117
    Log in for availability
    1 * 1 ST - 700,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Cleaning tissues
    Cleaning tissues
    METT51325003
    Log in for availability
    1 * 10 ST - 39,50 €