Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Bath fluids

Thermal HY, −80...+55 °C, 80 °C, 78 °C, Clear, 10 L
Catalog # 461-0131
Supplier:   JULABO GmbH
Bath fluids
Catalog # 461-0131
Supplier:  JULABO GmbH
CAS Number:  
Bath fluids
Bath fluids

Thermal HY, −80...+55 °C, 80 °C, 78 °C, Clear, 10 L

CAS Number:  
Price
820,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever water cannot be employed as bath fluid. The choice of bath fluid is critical for best results in temperature control and operation of the circulator.

  • Low toxicity and almost odourless
  • High stability and good thermal conductivity
  • Low viscosity and low corrosiveness
Specifications

Specifications

  • Pk:
    10 L
  • Colour:
    Clear
  • Fire point:
    80 °C
  • Flash point:
    78 °C
  • Temperature range:
    −80...+55 °C
  • Type:
    Thermal HY

Specifications