Orders LinkContactUsLinkComponent
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
90 mm
Catalog # 511-0838
Supplier:   DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
Catalog # 511-0838
Supplier:  DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
90 mm
Price
435,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.
Specifications
  • For filter Ø:
    90 mm