Orders LinkContactUsLinkComponent
Büchner funnels, Hartley pattern
70 mm
  511-0837
Büchner funnels, Hartley pattern
  511-0837
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
70 mm
342,00 €
1 * 1 ST
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.
  • For filter Ø:
    70 mm