Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
Separating funnels, pear shaped
100 ml, 19/26 NS
Catalog # 511-0754
Supplier:   SciLabware
Separating funnels, pear shaped
Catalog # 511-0754
Supplier:  SciLabware
Separating funnels, pear shaped
Separating funnels, pear shaped
100 ml, 19/26 NS
Price
102,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml