Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
2000 ml, 29/32 NS
Catalog # 511-0764
Supplier:   SciLabware
Conical separating funnels
Catalog # 511-0764
Supplier:  SciLabware
Conical separating funnels
Conical separating funnels
2000 ml, 29/32 NS
Price
190,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    2000 ml
  • Seq:
    06