Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
100 ml, 19/26 NS
Catalog # 511-0760
Supplier:   SciLabware
Conical separating funnels
Catalog # 511-0760
Supplier:  SciLabware
Conical separating funnels
Conical separating funnels
100 ml, 19/26 NS
Price
93,60 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.
Spezifikationen
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml
  • Seq:
    02