Orders LinkContactUsLinkComponent
Calibration weights, cylindrical, OIML, class E2
Calibration weights, cylindrical, OIML, class E2
5 g, 0,050 ± mg, Cylindrical test weight
Catalog # SARBYCW3528-02
Supplier:   Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Catalog # SARBYCW3528-02
Supplier:  Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Calibration weights, cylindrical, OIML, class E2
5 g, 0,050 ± mg, Cylindrical test weight
Price
186,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Class E2 cyclindrical calibration weights.
Specifications
  • Weight:
    5 g
  • Seq:
    15
  • Tolerance:
    0,050 ± mg
  • Type:
    Cylindrical test weight