Orders LinkContactUsLinkComponent
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
22×32 mm, #1
Catalog # 631-0133
Supplier:   VWR Collection
VWR® Rectangular Cover Glasses
Catalog # 631-0133
Supplier:  VWR Collection
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
22×32 mm, #1
Price
54,20 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
 : In plastic hinged lid boxes.
Specifications
  • No.:
    #1
  • L×W:
    22×32 mm
  • Seq:
    03