Orders LinkContactUsLinkComponent
Micro pipettes, ringcaps®
Micro pipettes, ringcaps®
50 µl, 125 mm, Standard
  612-2409
 :   HIRSCHMANN
Micro pipettes, ringcaps®
  612-2409
Micro pipettes, ringcaps®
Micro pipettes, ringcaps®
50 µl, 125 mm, Standard
28,60 €
1 * 250 ST
DURAN®, borosilicate glass, with ring mark, disposable.
  • Colour coded according to ISO for clear identification
  • Conformity certified
ISO 9000, DIN ISO 7550, IVD 98/79 EC
Certifications: ISO 9000, DIN ISO 7550, IVD 98/79 EC
Documents
  • Type:
    Standard
  • Capacity:
    50 µl
  • Length:
    125 mm