Orders LinkContactUsLinkComponent
Micro pipettes, ringcaps®
Micro pipettes, ringcaps®
5/10 µl, 125 mm, Standard
Catalog # 612-2402
Supplier:   HIRSCHMANN
Micro pipettes, ringcaps®
Catalog # 612-2402
Supplier:  HIRSCHMANN
Micro pipettes, ringcaps®
Micro pipettes, ringcaps®
5/10 µl, 125 mm, Standard
Price
30,60 €
Log in for availability
1 * 250 ST
Product Details & Documents
DURAN®, borosilicate glass, with ring mark, disposable.
  • Colour coded according to ISO for clear identification
  • Conformity certified
ISO 9000, DIN ISO 7550, IVD 98/79 EC
 : ISO 9000, DIN ISO 7550, IVD 98/79 EC
Documents
Specifications
  • Type:
    Standard
  • Capacity:
    5/10 µl
  • Length:
    125 mm