Order Entry
Austria
Orders LinkContactUsLinkComponent
Buchner funnels

Buchner funnels

40 mm, 80 ml, P40
Catalog # 511-0806
Supplier:   DIXON SCIENTIFIC
Buchner funnels
Catalog # 511-0806
Supplier:  DIXON SCIENTIFIC
Buchner funnels
Buchner funnels

40 mm, 80 ml, P40

Price
95,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass

  • With sintered glass disc
  • Very good chemical resistance
  • High temperature resistance

For paper or membrane filtration disc.

Specifications
  • Plate Ø:
    40 mm
  • Porosity:
    P40
  • Capacity:
    80 ml