Orders LinkContactUsLinkComponent
pH indicator papers, MQuant®, Supelco®
pH indicator papers, MQuant®, Supelco®
Special indicator, 5,4 - 7,0, 0,2 - 0,4
Catalog # 1.09556.0003
Supplier:   Merck
pH indicator papers, MQuant®, Supelco®
Catalog # 1.09556.0003
Supplier:  Merck
pH indicator papers, MQuant®, Supelco®
pH indicator papers, MQuant®, Supelco®
Special indicator, 5,4 - 7,0, 0,2 - 0,4
Price
56,10 €
Einloggen für Verfügbarkeit
1 * 3 Rolle
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
Spezifikationen
  • pH gradation:
    0,2 - 0,4
  • pH range:
    5,4 - 7,0
  • Description:
    Special indicator
  • Cat. No.:
    1.09556.0003
  • M-Clarity quality level:
    MQ100