Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Halogen bulbs

Halogen bulb, 12 V, 100 W
  ZEIS3800591660000
 :   ZEISS
Halogen bulbs
  ZEIS3800591660000
 :  ZEISS
Halogen bulbs

Halogen bulb, 12 V, 100 W

 
 
   
1 * 1 ST
 
 
  • Description:
    Halogen bulb
  • Seq:
    06
  • Power:
    100 W
  • Voltage:
    12 V