Orders LinkContactUsLinkComponent
Sponge swabs, Polywipe®
Sponge swabs, Polywipe®
Swab containers, 110 ml
Catalog # 720-0144
Sponge swabs, Polywipe®
Catalog # 720-0144
Sponge swabs, Polywipe®
Sponge swabs, Polywipe®
Swab containers, 110 ml
Price
192,00 €
Log in for availability
1 * 30 ST
Product Details & Documents
Non-inhibitory sponge material allows effective sampling of larger areas for environmental and hygiene monitoring. Available dry or pre-moistened with sterile diluent. Polywipes are supplied in peel pouches for immediate use, or in a 110 ml blue-capped container for transfer to the laboratory.
  • Blue colour for visibility
  • Triple wrapped and radiation sterilised
Delivery information: Supplied with individual sterile glove for handling.
Specifications
  • Description:
    Swab containers, 110 ml