Orders LinkContactUsLinkComponent
Inserts
E 134 Insert for 210 slides, equipped with 26 compartments at 26×11 mm, wire thickness 3 mm, W×D×H: 525×528×141 mm
Catalog # 141-0355
Supplier:   MIELE
Inserts
Catalog # 141-0355
Supplier:  MIELE
Inserts
Inserts
E 134 Insert for 210 slides, equipped with 26 compartments at 26×11 mm, wire thickness 3 mm, W×D×H: 525×528×141 mm
Price
349,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 134 Insert for 210 slides, equipped with 26 compartments at 26×11 mm, wire thickness 3 mm, W×D×H: 525×528×141 mm
  • Seq:
    48