Orders LinkContactUsLinkComponent
Inserts
Inserts
E 106 Insert for wide neck flasks and measuring cylinders, equipped with 10 spring hooks ×175 mm height and 16×105 mm height
Catalog # 141-0296
Supplier:   MIELE
Inserts
Catalog # 141-0296
Supplier:  MIELE
Inserts
Inserts
E 106 Insert for wide neck flasks and measuring cylinders, equipped with 10 spring hooks ×175 mm height and 16×105 mm height
Price
201,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 106 Insert for wide neck flasks and measuring cylinders, equipped with 10 spring hooks ×175 mm height and 16×105 mm height
  • Seq:
    31