Buffer solution pH 4.006 @25?C NIST and DIN 16266, 6x 250ml, Mettler 1 * 6 items
Catalog # METN51319004
Supplier: Mettler - Toledo
Buffer solution pH 4.006 @25?C NIST and DIN 16266, 6x 250ml, Mettler 1 * 6 items
Catalog # METN51319004
Supplier: Mettler - Toledo
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solution pH 6.865 @25?C NIST and DIN 19266, 6x250ml, Mettler 1 * 6 items
Supplier: Mettler - Toledo
Electrode cleaning solutions
Supplier: Mettler - Toledo